missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biochrom Sodium Regeneration Buffer 6
Biochrom Amino Acid Analysis reagents and buffers are manufactured from high quality chemicals and materials and subjected to rigorous control procedures, under the ISO 13485 Quality System.
Supplier: Biochrom 80203757
Description
Biochrom buffers are guaranteed to give accurate and reproducible results for up to 3 years from manufacture, provided the storage conditions are adhered to. All reagents have stable chemistry and have been formulated to be stored at ambient temperature so, conveniently, do not need refrigeration.Specifications
| Buffer | |
| Store at ambient temperature | |
| 1 L | |
| Liquid |