Learn More
Sodium Pyrophosphate Decahydrate, ACS Grade, Fisher Chemical™
Supplier: Thermo Fisher Scientific S39010
Specifications
| 93.8°C | |
| 80°C | |
| White | |
| Solid | |
| 99 to 103 % | |
| MFCD00149200 | |
| VZWGHDYJGOMEKT-UHFFFAOYSA-J | |
| tetrasodium;phosphonato phosphate;decahydrate | |
| 3084150 | |
| 99 to 103% | |
| Pass Test | |
| >94°C |
| Sodium Pyrophosphate Decahydrate | |
| 13472-36-1 | |
| 10 | |
| 10 kg | |
| H20Na4O17P2 | |
| sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate | |
| O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+] | |
| 446.05 | |
| 446.06 | |
| ACS | |
| Poly Pail |
Chemical Identifiers
| 13472-36-1 | |
| 446.05 | |
| VZWGHDYJGOMEKT-UHFFFAOYSA-J | |
| 3084150 | |
| O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+] |
| H20Na4O17P2 | |
| MFCD00149200 | |
| sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate | |
| tetrasodium;phosphonato phosphate;decahydrate |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . .
NFPA
Health:2
Flammability:1
Instability:0