missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Pyrophosphate Decahydrate, ACS Grade, Fisher Chemical™
Specifications
Specifications
| Boiling Point | 93.8°C |
| Melting Point | 80°C |
| Color | White |
| pH | 10 |
| Physical Form | Solid |
| Quantity | 500 g |
| Assay Percent Range | 99 to 103 % |
| Formula Weight | 446.06 |
| Percent Purity | 99 to 103% |
| Grade | ACS |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . . NFPA Health:2 Flammability:1 Instability:0 |