missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium pyrophosphate, 200mM buffer soln.

CAS: 7758-16-9

$154.95 - $255.92

Chemical Identifiers

CAS 7758-16-9
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62052AE
View Documents
Thermo Scientific Chemicals
J62052AE
100 mL
Each for $154.95
Only null left
 
AAJ62052AK
View Documents
Thermo Scientific Chemicals
J62052AK
250 mL
Each for $255.92
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium pyrophosphate, 200mM buffer soln. is used in the laboratory as a buffering agent. DSPP has been used as a chelating agent in antimicrobial studies. Tetrabasic pyrophosphate or TSPP (sc-203404; sc-236921) has shown to be useful in the preparation of an EDTA-sodium pyrophosphate extraction buffer for microcystin analysis of soil samples. It is a popular leavening agent found in baking powders.

Solubility
Fully miscible in water. Insoluble in alcohol and ammonia.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

7758-16-9

Specifications

7758-16-9
200mM buffer soln.
Liquid
100 mL
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient Temperatures

SDS
Documents

Documents

RUO – Research Use Only