accessibility menu, dialog, popup

UserName

NaNaNaNaOPOOOPOOO

CAS RN 7758-16-9

IUPAC Name: tetrasodium (phosphonatooxy)phosphonate
Synonyms: tetrasodium pyrophosphate disodium pyrophosphate
Molecular Weight (g/mol): 265.9
Molecular Formula: Na4O7P2
InChi Key: FQENQNTWSFEDLI-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O
Physical Form 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)
Percent Purity 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (2)