missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium pyrophosphate, 200mM buffer soln.

Catalog No. AAJ62052AK
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62052AK 250 mL
AAJ62052AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62052AK Supplier Thermo Scientific Chemicals Supplier No. J62052AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7758-16-9

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium pyrophosphate, 200mM buffer soln. is used in the laboratory as a buffering agent. DSPP has been used as a chelating agent in antimicrobial studies. Tetrabasic pyrophosphate or TSPP (sc-203404; sc-236921) has shown to be useful in the preparation of an EDTA-sodium pyrophosphate extraction buffer for microcystin analysis of soil samples. It is a popular leavening agent found in baking powders.

Solubility
Fully miscible in water. Insoluble in alcohol and ammonia.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 7758-16-9

Specifications

Concentration 200mM buffer soln.
Physical Form Liquid
Quantity 250 mL
Recommended Storage Ambient Temperatures

RUO – Research Use Only