missing translation for 'onlineSavingsMsg'
Learn More

Sodium Propionate 98.0+%, TCI America™

Supplier:  TCI America P0512500G

 View more versions of this product

Catalog No. P0512500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Propionate
White
7.0 to 10.0 (50g/L 25°C)
500 g
MFCD00002759
JXKPEJDQGNYQSM-UHFFFAOYSA-M
sodium;propanoate
2723816
≥98.0% (T)
286°C
137-40-6
Crystalline Powder
C3H5NaO2
sodium propionate, sodium propanoate, propionic acid sodium salt, propanoic acid, sodium salt, napropion, ocuseptine, deketon, impedex, keenate, propiofar
CCC(=O)[O-].[Na+]
96.061
96.06

Chemical Identifiers

137-40-6
96.061
JXKPEJDQGNYQSM-UHFFFAOYSA-M
2723816
CCC(=O)[O-].[Na+]
C3H5NaO2
MFCD00002759
sodium propionate, sodium propanoate, propionic acid sodium salt, propanoic acid, sodium salt, napropion, ocuseptine, deketon, impedex, keenate, propiofar
sodium;propanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1203

RTECSNumber : UF7525000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions