missing translation for 'onlineSavingsMsg'
Learn More

Sodium Propionate 98.0+%, TCI America™

Catalog No. P051225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P051225G 25 g
P0512500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P051225G Supplier TCI America Supplier No. P051225G

Chemical Identifiers

CAS 137-40-6
Molecular Formula C3H5NaO2
Molecular Weight (g/mol) 96.061
MDL Number MFCD00002759
InChI Key JXKPEJDQGNYQSM-UHFFFAOYSA-M
Synonym sodium propionate, sodium propanoate, propionic acid sodium salt, propanoic acid, sodium salt, napropion, ocuseptine, deketon, impedex, keenate, propiofar
PubChem CID 2723816
IUPAC Name sodium;propanoate
SMILES CCC(=O)[O-].[Na+]

Specifications

Melting Point 286°C
Color White
pH 7.0 to 10.0 (50g/L 25°C)
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 96.06
Percent Purity ≥98.0% (T)
Chemical Name or Material Sodium Propionate
EINECSNumber (9)-1203
RTECSNumber UF7525000
TSCA Yes