missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Propionate, NF, 99-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Sodium Propionate, NF is used as an antiseptic agent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Fiber Drum |
| Quantity | 45 kg |
| Percent Purity | 99 to 100.5% |
| Grade | NF |