accessibility menu, dialog, popup

UserName

H2ONaH3COO

CAS RN 6700-17-0

IUPAC Name: sodium hydrate propanoate
Synonyms: sodium hydrate propionate
Molecular Weight (g/mol): 114.08
Molecular Formula: C3H7NaO3
InChi Key: HOAUAOBUGFYWMK-UHFFFAOYSA-M
SMILES: O.[Na+].CCC([O-])=O
Grade 
  • (4)
Percent Purity 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results