missing translation for 'onlineSavingsMsg'
Learn More

Sodium Propionate, FCC, 99-100.5%, Spectrum™ Chemical

SO192, 137-40-6, C3H5NaO2

Supplier:  Spectrum Chemical Mfg Cor SO19225KG

 View more versions of this product

Catalog No. 18606757


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Sodium Propionate, FCC is used primarily as a food preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

137-40-6
Poly Pail
2.5 kg
[Na+].CCC([O-])=O
96.06
FCC
1
C3H5NaO2
JXKPEJDQGNYQSM-UHFFFAOYSA-M
sodium propanoate
99 to 100.5%

Chemical Identifiers

137-40-6
96.06
sodium propanoate
C3H5NaO2
JXKPEJDQGNYQSM-UHFFFAOYSA-M
[Na+].CCC([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions