missing translation for 'onlineSavingsMsg'
Learn More

Sodium Propionate, FCC, 99-100.5%, Spectrum™ Chemical

Catalog No. 18606756
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606756 500 g Poly Pail
18606757 2.5 kg Poly Pail
18606758 12 kg Fiber Drum
18606759 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606756 Supplier Spectrum Chemical Mfg Cor Supplier No. SO192500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

SO192, 137-40-6, C3H5NaO2

Spectrum™ Chemical Sodium Propionate, FCC is used primarily as a food preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 137-40-6
Molecular Formula C3H5NaO2
Molecular Weight (g/mol) 96.06
InChI Key JXKPEJDQGNYQSM-UHFFFAOYSA-M
IUPAC Name sodium propanoate
SMILES [Na+].CCC([O-])=O

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 500 g
Percent Purity 99 to 100.5%
Grade FCC