missing translation for 'onlineSavingsMsg'
Learn More

Sodium Potassium Tartrate, 25% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company 755016

Quantity 500 mL
Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. 755016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

6381-59-5 , 7732-18-5
282.218
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
C4H12KNaO10
VZOPRCCTKLAGPN-UHFFFAOYSA-L
2724148
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]

Specifications

Sodium Potassium Tartrate
78.0, 19.61
C4H12KNaO10
VZOPRCCTKLAGPN-UHFFFAOYSA-L
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
2724148
Aqueous Solution
25% (w/v)
6381-59-5 , 7732-18-5
82.0, 20.41
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
282.218
Natural Poly Bottle
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents