missing translation for 'onlineSavingsMsg'
Learn More

Sodium Polytungstate Solution, ∼85% in H2O, For separation of minerals, MilliporeSigma™ Supelco™

$936.71 - $3,773.55

Specifications

CAS 12141-67-2
Linear Formula Na6O39W12
MDL Number MFCD00061466
Formula Weight 2968.00
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111012770
View Documents
Merck Emd Millipore
80656100ML
100 mL N/A
Only null left
 
111012771
View Documents
Merck Emd Millipore
80656500ML
500 mL N/A
Only null left
 
Description

Description

Specifications

Specifications

12141-67-2
Na6O39W12
MFCD00061466
2968.00
Videos
SDS
Documents

Documents