missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Polytungstate Solution, ∼85% in H2O, For separation of minerals, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Hexasodium Dihydrogen-dodecawolframate Monohydrate |
| CAS | 12141-67-2 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Linear Formula | Na6O39W12 |
| MDL Number | MFCD00061466 |
| Molecular Weight (g/mol) | 2968.00 |
| Formula Weight | 2968.00 |
| Grade | Reagent |