missing translation for 'onlineSavingsMsg'
Learn More

Sodium polyanetholesulfonate

Catalog No. AC419635000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC419635000 500 g
AC419630250 25 g
AC419631000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC419635000 Supplier Thermo Scientific Chemicals Supplier No. 419635000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55963-78-5 | (C10H11NaO4S)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55963-78-5
Molecular Formula (C10H11NaO4S)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00148427
InChI Key JKJBFNAERWARKW-CZEFNJPISA-L
Synonym sodium anethole sulfite, polyanetholesulfonate, polyanethol sulfonate, benzene, 1-methoxy-4-1-propenyl-, e-, homopolymer, sulfonated, sodium salt, sodium 1-methoxy-4-e-prop-1-enyl benzene sulfite
PubChem CID 6434512
SMILES CS(=O)(=O)O[Na].COC1=CC=C(C=C1)C(-*)C(C)-*

Specifications

Color Tan to White
pH 5.0 to 7.0 (1% aq. soln.)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500 g
Merck Index 14,8666
Solubility Information Solubility in water: soluble.
IUPAC Name sodium;1-methoxy-4-[(E)-prop-1-enyl]benzene;sulfite
Physical Form Crystals or Powder
Chemical Name or Material Sodium polyanetholesulfonate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only