missing translation for 'onlineSavingsMsg'
Learn More

Sodium, plasma standard solution, Specpure™ Na 10,000μg/mL

Na2CO3 in 5% HNO3

$149.52 - $527.82

Chemical Identifiers

Molecular Formula Na2CO3 in 5% HNO3
MDL Number MFCD00085307
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA14400AD
View Documents
Thermo Scientific Chemicals
014400AD
50 mL N/A
Only null left
 
AA14400AE
View Documents
Thermo Scientific Chemicals
014400AE
100 mL N/A
Only null left
 
AA14400AP
View Documents
Thermo Scientific Chemicals
014400AP
500 mL N/A
Only null left
 
Description

Description

Sodium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

Na2CO3 in 5% HNO3
MFCD00085307

Specifications

23 hPa (17mm Hg) at 20°C
Na2CO3 in 5% HNO3
Miscible with water
Nitric acid: 5%; Sodium carbonate: 2.3%; Water: 92.7%
Liquid
MFCD00085307
50 mL
Sodium, plasma standard solution
Videos
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 231-714-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only