missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pivalate Hydrate 98.0+%, TCI America™

Catalog No. p-7136009
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09785G 5 g
S097825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S09785G Supplier TCI America Supplier No. S09785G
Only null left

Chemical Identifiers

CAS 143174-36-1
Molecular Formula C5H9NaO2
Molecular Weight (g/mol) 124.12
MDL Number MFCD00150782
InChI Key SJRDNQOIQZOVQD-UHFFFAOYSA-M
Synonym sodium trimethylacetate hydrate, sodium pivalate hydrate, sodium hydrate pivalate, propanoic acid, 2,2-dimethyl-, sodium salt, hydrate, acmc-20ajn9, sodiumtrimethylacetatehydrate, c5h9o2.na.h2o, trimethylacetic acid sodium salt hydrate, sodium 2,2-dimethylpropanoate-water 1/1/1
PubChem CID 23666172
IUPAC Name sodium 2,2-dimethylpropanoate
SMILES [Na+].CC(C)(C)C([O-])=O

Specifications

Color White
Quantity 5 g
Formula Weight 124.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Pivalate Hydrate
RTECSNumber UA2459665
TSCA No