accessibility menu, dialog, popup

UserName

CAS RN 143174-36-1

IUPAC Name: sodium 2,2-dimethylpropanoate
Synonyms: sodium pivalate
Molecular Weight (g/mol): 124.12
Molecular Formula: C5H9NaO2
InChi Key: SJRDNQOIQZOVQD-UHFFFAOYSA-M
SMILES: [Na+].CC(C)(C)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results