missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Tribasic Dodecahydrate, Honeywell Fluka™

Catalog No. p-7071900
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002141 500 g
6002140 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002141 Supplier Honeywell Chemicals Supplier No. 222003500G
Only null left

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10101-89-0
Molecular Formula H24Na3O16P
Molecular Weight (g/mol) 380.119
MDL Number MFCD00149198
InChI Key ASTWEMOBIXQPPV-UHFFFAOYSA-K
Synonym trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate
PubChem CID 61473
IUPAC Name trisodium;phosphate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Quantity 500 g
Linear Formula Na3PO4 · 12H2O
UN Number NONH for all modes of transport
Formula Weight 380.12g/mol
Percent Purity ≥98%
Grade ACS Reagent
Density 1.62g/mL at 25°C
Chemical Name or Material Sodium Phosphate Tribasic Dodecahydrate
Hazard Statement P305 + P351 + P338
Precautionary Statement H315-H319-H335
EINECSNumber 231-509-8
RTECSNumber TC9575000