missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sodium phosphate, tribasic dodecahydrate, 98%, for biochemistry

Supplier:  Thermo Scientific™ 447965000

 View more versions of this product

Catalog No. AC447965000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Biological buffer
Specifications

Specifications

Sodium phosphate, tribasic dodecahydrate
500g
Na3O412H2O
15,8792
Solubility in water: 240g/L (20°C). Other solubilities: practically insoluble in alcohol
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]
380.13
380.13
Biochemistry
1.62
10101-89-0
98%
Na3PO4·12H2O
trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate
ASTWEMOBIXQPPV-UHFFFAOYSA-K
trisodium;phosphate;dodecahydrate
61473
98%
Plastic bottle
1.6200g/mL

Chemical Identifiers

10101-89-0
380.13
trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate
trisodium;phosphate;dodecahydrate
Na3O412H2O
ASTWEMOBIXQPPV-UHFFFAOYSA-K
61473
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only