missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Monobasic, MP Biomedicals
Supplier: MP Biomedicals Inc 0219550090
Specifications
| Sodium Phosphate Monobasic | |
| 7558-80-7 | |
| H2NaO4P | |
| monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
| [Na+].OP(O)([O-])=O | |
| 119.98 | |
| CHEBI:37585 |
| Anhydrous | |
| 500 g | |
| MFCD00003527,MFCD00146206 | |
| AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
| sodium dihydrogen phosphate | |
| 23672064 | |
| 120 |
Chemical Identifiers
| 7558-80-7 | |
| 119.98 | |
| AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
| 23672064 | |
| sodium dihydrogen phosphate |
| H2NaO4P | |
| MFCD00003527,MFCD00146206 | |
| monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
| CHEBI:37585 | |
| [Na+].OP(O)([O-])=O |
Safety and Handling
Recommended Storage : Room Temperature