missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Monobasic, MP Biomedicals
Specifications
Specifications
| Quantity | 3 kg |
| Formula Weight | 120 |
| Chemical Name or Material | Sodium Phosphate Monobasic, Anhydrous |
Safety and Handling
| Recommended Storage | Room Temperature |