missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Monobasic, MP Biomedicals

Catalog No. ICN19550080
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19550080 100 g
ICN19550090 500 g
ICN19550091 1 kg
ICN19550092 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19550080 Supplier MP Biomedicals Inc Supplier No. 0219550080
Only null left

Chemical Identifiers

CAS 7558-80-7
Molecular Formula H2NaO4P
Molecular Weight (g/mol) 119.98
MDL Number MFCD00003527,MFCD00146206
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
Synonym monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate
PubChem CID 23672064
ChEBI CHEBI:37585
IUPAC Name sodium dihydrogen phosphate
SMILES [Na+].OP(O)([O-])=O

Specifications

Quantity 100 g
Formula Weight 120
Chemical Name or Material Sodium Phosphate Monobasic, Anhydrous
Recommended Storage Room Temperature