missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate monobasic monohydrate, puriss. p.a., ACS Reagent, Honeywell Fluka™

Catalog No. p-7072258
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002841 250 g
6002840 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002841 Supplier Honeywell Chemicals Supplier No. 71504250G
Only null left

Puriss. p.a., ACS Reagent,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
IUPAC Name sodium;dihydrogen phosphate;hydrate
SMILES O.[Na+].OP(O)(O)=O

Specifications

Quantity 250 g
Linear Formula NaH2PO4 · H2O
UN Number NONH for all modes of transport
Formula Weight 137.99g/mol
Percent Purity ≥99% (T)
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium phosphate monobasic monohydrate