missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Sodium Phosphate, Monobasic, Monohydrate, OmniPur™, Calbiochem™,
$156.10 - $156.10
Chemical Identifiers
| CAS | 10049-21-5 |
|---|---|
| Molecular Formula | H5NaO5P |
| Molecular Weight (g/mol) | 139.00 |
| MDL Number | MFCD00149208 |
| InChI Key | BBMHARZCALWXSL-UHFFFAOYSA-N |
| Synonym | sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 |
| PubChem CID | 516949 |
| IUPAC Name | sodium phosphoric acid hydrate |
| SMILES | O.[Na+].OP(O)(O)=O |
Chemical Identifiers
| 10049-21-5 | |
| 139.00 | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| 516949 | |
| O.[Na+].OP(O)(O)=O |
| H5NaO5P | |
| MFCD00149208 | |
| sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
| sodium phosphoric acid hydrate |
Specifications
| 10049-21-5 | |
| 98.0 to 102.0% | |
| MFCD00149208 | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| sodium phosphoric acid hydrate | |
| 516949 | |
| Sodium Phosphate, Monobasic, Monohydrate |
| 500 g | |
| H5NaO5P | |
| sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
| O.[Na+].OP(O)(O)=O | |
| 139.00 | |
| 137.99 |