missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, monobasic monohydrate, 98%, extra pure

Catalog No. AC447740010
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC447740010 1 kg
AC447740050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447740010 Supplier Thermo Scientific Chemicals Supplier No. 447740010

Sodium phosphate, monobasic monohydrate, 98%, H2NaO4P.H2O, CAS Number-10049-21-5 | CAS: 10049-21-5 | H5NaO5P | 139.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
SMILES O.[Na+].OP(O)(O)=O

Specifications

Melting Point 100.0°C
Quantity 1 kg
Assay Percent Range 98%
Linear Formula NaH2PO4·H2O
Merck Index 15,879
Solubility Information Solubility in water: 1103g/L (20°C). Other solubilities: practically insoluble in alcohol
IUPAC Name sodium;dihydrogen phosphate;hydrate
Formula Weight 137.99
Percent Purity 98%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Sodium phosphate, monobasic monohydrate
Show More Show Less

RUO – Research Use Only