missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Monobasic Monohydrate, 98.0-102.0%, (White crystalline powder/ACS Reagent Grade), MP Biomedicals
$338.16 - $1131.36
Chemical Identifiers
| CAS | 10049-21-5 |
|---|---|
| Molecular Formula | H5NaO5P |
| Molecular Weight (g/mol) | 139.00 |
| MDL Number | MFCD00149208 |
| InChI Key | BBMHARZCALWXSL-UHFFFAOYSA-N |
| Synonym | sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 |
| PubChem CID | 516949 |
| IUPAC Name | sodium phosphoric acid hydrate |
| SMILES | O.[Na+].OP(O)(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19144201
|
MP Biomedicals Inc
0219144201 |
1 kg |
Each for $338.16
|
|
|||||
|
ICN19144205
|
MP Biomedicals Inc
0219144205 |
5 kg |
Each for $1,131.36
|
|
|||||
Chemical Identifiers
| 10049-21-5 | |
| 139.00 | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| 516949 | |
| O.[Na+].OP(O)(O)=O |
| H5NaO5P | |
| MFCD00149208 | |
| sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
| sodium phosphoric acid hydrate |
Specifications
| White | |
| Crystalline Powder | |
| 98.0-102.0% | |
| MFCD00149208 | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| sodium phosphoric acid hydrate | |
| 516949 | |
| ACS Reagent |
| 10049-21-5 | |
| 1 kg | |
| H5NaO5P | |
| sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
| O.[Na+].OP(O)(O)=O | |
| 139.00 | |
| 137.99 | |
| Sodium Phosphate Monobasic Monohydrate |
Safety and Handling
Recommended Storage : Room Temperature