missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate monobasic, Honeywell Fluka™

Catalog No. p-7072257
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
Catalog No. Quantity
6002838 100 g
1 options

Catalog No. 6002838

Supplier: Honeywell Chemicals 71492100G

Only null left

TraceSELECT™, for trace analysis, anhydrous,≥99.999% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

Chemical Identifiers

CAS 7558-80-7
Molecular Formula H2NaO4P
Molecular Weight (g/mol) 119.98
MDL Number MFCD00003527,MFCD00146206
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
Synonym monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate
PubChem CID 23672064
ChEBI CHEBI:37585
IUPAC Name sodium;dihydrogen phosphate
SMILES [Na+].OP(O)([O-])=O

Specifications

Quantity 100 g
Linear Formula NaH2PO4
UN Number NONH for all modes of transport
Formula Weight 119.98g/mol
Percent Purity ≥99.99% (metals basis)
Chemical Name or Material Sodium phosphate monobasic
RTECSNumber WA1900000