missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate monobasic, For HPLC, ≥99.0%, Honeywell Fluka™

Catalog No. p-7122047
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60044959 50 g
6002037 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60044959 Supplier Honeywell Chemicals Supplier No. 1784450G
Only null left

for HPLC,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7558-80-7
Molecular Formula H2NaO4P
Molecular Weight (g/mol) 119.98
MDL Number MFCD00003527,MFCD00146206
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
Synonym monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate
PubChem CID 23672064
ChEBI CHEBI:37585
IUPAC Name sodium;dihydrogen phosphate
SMILES [Na+].OP(O)([O-])=O

Specifications

CAS Max % 100.0000%
Quantity 50 g
Linear Formula NaH2PO4
UN Number NONH for all modes of transport
Solubility Information Soluble in water 0.1g/mL at 25°C
Formula Weight 119.98
Percent Purity ≥99%
Grade HPLC
Packaging Glass Bottle
Density 2.36 g/cm3
Chemical Name or Material Sodium phosphate monobasic, For HPLC, ≥99.0%
Show More Show Less
ShelfLife 1620 days from date of manufacture
EINECSNumber 231-449-2
RTECSNumber WA1900000
Recommended Storage Room Temp