missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate monobasic, For HPLC, ≥99.0%, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 250 g |
| Linear Formula | NaH2PO4 |
| UN Number | NONH for all modes of transport |
| Solubility Information | Soluble in water 0.1g/mL at 25°C |
| Formula Weight | 119.98g/mol |
| Percent Purity | ≥99% |
| Packaging | Glass bottle |
| Chemical Name or Material | Sodium Phosphate Monobasic |
Safety and Handling
| EINECSNumber | 231-449-2 |
| RTECSNumber | WA1900000 |