missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Monobasic Dihydrate (Crystalline/Certified), Fisher Chemical™

Supplier:  Thermo Fisher Scientific S3813

 View more versions of this product

Catalog No. S381-3

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Phosphate Monobasic Dihydrate
White
Solid
≥99 %
MFCD00149209
VBJGJHBYWREJQD-UHFFFAOYSA-M
sodium;dihydrogen phosphate;dihydrate
23673460
≥99%
Pass Test
Poly Bottle
225°C
13472-35-0
3 kg
H6NaO6P
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
O.O.OP(=O)(O)[O-].[Na+]
156.006
156.01
Certified
21 to 25%

Chemical Identifiers

13472-35-0
156.006
VBJGJHBYWREJQD-UHFFFAOYSA-M
23673460
O.O.OP(=O)(O)[O-].[Na+]
H6NaO6P
MFCD00149209
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
sodium;dihydrogen phosphate;dihydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions