missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Monobasic Dihydrate (Crystalline/Certified), Fisher Chemical™
$350.15 - $1008.31
Chemical Identifiers
| CAS | 13472-35-0 |
|---|---|
| Molecular Formula | H6NaO6P |
| Molecular Weight (g/mol) | 156.006 |
| MDL Number | MFCD00149209 |
| InChI Key | VBJGJHBYWREJQD-UHFFFAOYSA-M |
| Synonym | sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh |
| PubChem CID | 23673460 |
| IUPAC Name | sodium;dihydrogen phosphate;dihydrate |
| SMILES | O.O.OP(=O)(O)[O-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
S381-500
|
Thermo Fisher Scientific
S381500 |
500 g | Poly Bottle |
Each for $350.15
Case of 6 Each for $2,100.90
|
|
||||
|
S381-3
|
Thermo Fisher Scientific
S3813 |
3 kg | Poly Bottle |
Each for $1,008.31
Case of 4 Each for $4,033.24
|
|
||||
Chemical Identifiers
| 13472-35-0 | |
| 156.006 | |
| VBJGJHBYWREJQD-UHFFFAOYSA-M | |
| 23673460 | |
| O.O.OP(=O)(O)[O-].[Na+] |
| H6NaO6P | |
| MFCD00149209 | |
| sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh | |
| sodium;dihydrogen phosphate;dihydrate |
Specifications
| 225°C | |
| 13472-35-0 | |
| 500 g | |
| H6NaO6P | |
| sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh | |
| O.O.OP(=O)(O)[O-].[Na+] | |
| 156.006 | |
| 156.01 | |
| Certified | |
| 21 to 25% | |
| Sodium Phosphate Monobasic Dihydrate |
| White | |
| Solid | |
| ≥99 % | |
| MFCD00149209 | |
| VBJGJHBYWREJQD-UHFFFAOYSA-M | |
| sodium;dihydrogen phosphate;dihydrate | |
| 23673460 | |
| ≥99% | |
| Pass Test | |
| Poly Bottle |
Safety and Handling