missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Monobasic Anhydrous (USP), Fisher Chemical

Catalog No. s397500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S397500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S397500 Supplier Thermo Fisher Scientific Supplier No. S397500
Only null left

Chemical Identifiers

CAS 7558-80-7
Molecular Formula H2NaO4P
Molecular Weight (g/mol) 119.98
MDL Number MFCD00003527,MFCD00146206
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
Synonym monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate
PubChem CID 23672064
ChEBI CHEBI:37585
IUPAC Name sodium dihydrogen phosphate
SMILES [Na+].OP(O)([O-])=O

Specifications

Boiling Point 203.9°C
Melting Point 100°C
Color White
pH 4.1 to 4.5
Physical Form Solid
Quantity 500 g
Assay Percent Range 98.0 to 103.0 %
Formula Weight 119.96
Percent Purity 98.0 to 103.0%
Grade USP
Identification Pass Test
Packaging Poly Bottle
Density 2.04g/cm³
Chemical Name or Material Sodium Phosphate Monobasic Anhydrous
Show More Show Less