missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Monobasic Anhydrous (Colorless-to-White Crystals), Fisher BioReagents™
Description
For tissue culture and molecular biology applications
Sodium Phosphate is commonly used in tissue culture and molecular biology applications.
Specifications
Specifications
| Boiling Point | 203.9°C |
| Melting Point | 100°C |
| Color | White |
| pH | 4.1 to 4.5 |
| Physical Form | Solid |
| Quantity | 500 g |
| Assay Percent Range | ≥99 % |
| Merck Index | 15, 8790 |
| IUPAC Name | sodium;dihydrogen phosphate |
| Formula Weight | 119.98 |
| Show More |
Safety and Handling
| Recommended Storage | RT |