missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Sodium Phosphate, Monobasic, ACS Reagent Grade, Calbiochem™,

Supplier:  MilliporeSigma™ 567545500GM

 View more versions of this product

Catalog No. 567545500GM


Only null left

Description

Description

Specifications

Specifications

White
500 g
H5NaO5P
NaH2PO4
O.[Na+].OP(O)(O)=O
139.00
≥98%
10049-21-5
Glass Bottle
MFCD00149208
BBMHARZCALWXSL-UHFFFAOYSA-N
sodium phosphoric acid hydrate
516949
ACS Reagent

Chemical Identifiers

10049-21-5
139.00
BBMHARZCALWXSL-UHFFFAOYSA-N
516949
O.[Na+].OP(O)(O)=O
H5NaO5P
MFCD00149208
NaH2PO4
sodium phosphoric acid hydrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions