missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Sodium Phosphate, Monobasic, ACS Reagent Grade, Calbiochem™,
Supplier: MilliporeSigma™ 5675451KG
| Quantity | 1 kg |
|---|---|
| Packaging | Glass Bottle |
Chemical Identifiers
| 10049-21-5 | |
| 139.00 | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| 516949 | |
| O.[Na+].OP(O)(O)=O |
| H5NaO5P | |
| MFCD00149208 | |
| NaH2PO4 | |
| sodium phosphoric acid hydrate |
Specifications
| White | |
| 1 kg | |
| H5NaO5P | |
| NaH2PO4 | |
| O.[Na+].OP(O)(O)=O | |
| 139.00 | |
| ≥98% |
| 10049-21-5 | |
| Glass Bottle | |
| MFCD00149208 | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| sodium phosphoric acid hydrate | |
| 516949 | |
| ACS Reagent |