missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium phosphate, monobasic, 99%, for biochemistry, anhydrous

Catalog No. AC447982500
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447982500 250 g Plastic bottle
AC447980010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447982500 Supplier Thermo Scientific Chemicals Supplier No. 447982500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7558-80-7 ∣ H2NaO4P ∣ 119.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7558-80-7
Molecular Formula H2NaO4P
Molecular Weight (g/mol) 119.98
MDL Number MFCD00003527,MFCD00146206
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
Synonym monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate
PubChem CID 23672064
ChEBI CHEBI:37585
IUPAC Name sodium dihydrogen phosphate
SMILES [Na+].OP(O)([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 99%
Linear Formula NaH2PO4
Merck Index 15,879
Solubility Information (1 M in water) Clear colorless
Formula Weight 119.98
Percent Purity 99%
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Sodium phosphate, monobasic
Show More Show Less
EINECSNumber 231-449-2

RUO – Research Use Only