missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate, monobasic, 98%, extra pure, anhydrous
CAS: 7558-80-7 | H2NaO4P | 119.98 g/mol
$136.37 - $136.37
Chemical Identifiers
| CAS | 7558-80-7 |
|---|---|
| Molecular Formula | H2NaO4P |
| Molecular Weight (g/mol) | 119.98 |
| MDL Number | MFCD00003527,MFCD00146206 |
| InChI Key | AJPJDKMHJJGVTQ-UHFFFAOYSA-M |
| Synonym | monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate |
| PubChem CID | 23672064 |
| ChEBI | CHEBI:37585 |
| IUPAC Name | sodium;dihydrogen phosphate |
| SMILES | [Na+].OP(O)([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC448170010
|
Thermo Scientific Chemicals
448170010 |
1 kg | Plastic bottle |
Each for $136.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7558-80-7 | |
| 119.98 | |
| AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
| 23672064 | |
| sodium;dihydrogen phosphate |
| H2NaO4P | |
| MFCD00003527,MFCD00146206 | |
| monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
| CHEBI:37585 | |
| [Na+].OP(O)([O-])=O |
Specifications
| 7558-80-7 | |
| 98% | |
| NaH2PO4 | |
| 15,879 | |
| Solubility in water: 850g/L (20°C). | |
| [Na+].OP(O)([O-])=O | |
| 119.98 | |
| CHEBI:37585 | |
| 98% | |
| Plastic bottle |
| 1 kg | |
| H2NaO4P | |
| MFCD00003527,MFCD00146206 | |
| monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
| AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
| sodium;dihydrogen phosphate | |
| 23672064 | |
| 119.98 | |
| Extra Pure | |
| Sodium phosphate, monobasic |
Safety and Handling
EINECSNumber : 231-449-2
RUO – Research Use Only