missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic, puriss p.a. ACS Reagent, Honeywell Fluka™
Puriss. p.a., ACS Reagent, anhydrous,≥99.0% (T)
$348.66 - $348.66
Chemical Identifiers
| CAS | 7558-79-4 |
|---|---|
| Molecular Formula | HNa2O4P |
| Molecular Weight (g/mol) | 141.96 |
| MDL Number | MFCD00003496 |
| InChI Key | BNIILDVGGAEEIG-UHFFFAOYSA-L |
| Synonym | disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt |
| PubChem CID | 24203 |
| ChEBI | CHEBI:34683 |
| IUPAC Name | disodium;hydrogen phosphate |
| SMILES | [Na+].[Na+].OP([O-])([O-])=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7558-79-4 | |
| 141.96 | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| 24203 | |
| disodium;hydrogen phosphate |
| HNa2O4P | |
| MFCD00003496 | |
| disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
| CHEBI:34683 | |
| [Na+].[Na+].OP([O-])([O-])=O |
Specifications
| 7558-79-4 | |
| HNa2O4P | |
| MFCD00003496 | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| disodium;hydrogen phosphate | |
| 24203 | |
| 141.96g/mol | |
| ACS/puriss. p.a. |
| 1 kg | |
| Na2HPO4 | |
| disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
| [Na+].[Na+].OP([O-])([O-])=O | |
| 141.96 | |
| CHEBI:34683 | |
| ≥99% (T) | |
| Sodium phosphate dibasic |
Safety and Handling
RTECSNumber : WC4500000