missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Dibasic, MP Biomedicals
Specifications
Specifications
| Quantity | 500 g |
| Formula Weight | 141.96 |
| Grade | Molecular Biology |
| Chemical Name or Material | Sodium Phosphate Dibasic, Anhydrous |