missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Heptahydrate (Crystalline/Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific S3733

 View more versions of this product

Catalog No. S373-3



Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Phosphate Dibasic Heptahydrate
White
9.0 to 9.3
3 kg
H15Na2O11P
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
O.[Na+].[Na+].OP([O-])([O-])=O
268.06
268.07
Certified ACS
Poly Bottle
48°C
7782-85-6
Solid
98 to 102 %
MFCD00149180
LVXHNCUCBXIIPE-UHFFFAOYSA-L
disodium phosphoric acid heptahydrate
6096963
98 to 102%
Pass Test

Chemical Identifiers

7782-85-6
268.06
LVXHNCUCBXIIPE-UHFFFAOYSA-L
6096963
O.[Na+].[Na+].OP([O-])([O-])=O
H15Na2O11P
MFCD00149180
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
disodium phosphoric acid heptahydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions