missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Heptahydrate (Crystalline/Certified ACS), Fisher Chemical™

$332.33 - $1,092.08

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H15Na2O11P
Molecular Weight (g/mol) 268.06
MDL Number MFCD00149180
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
PubChem CID 6096963
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.[Na+].[Na+].OP([O-])([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
S373-500
SDS View Documents Promotion Details
Thermo Fisher Scientific
S373500
500 g Poly Bottle N/A
Only null left
 
S373-3
SDS View Documents Promotion Details
Thermo Fisher Scientific
S3733
3 kg Poly Bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7782-85-6
268.06
LVXHNCUCBXIIPE-UHFFFAOYSA-L
6096963
O.[Na+].[Na+].OP([O-])([O-])=O
H15Na2O11P
MFCD00149180
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
disodium phosphoric acid heptahydrate

Specifications

48°C
7782-85-6
Solid
98 to 102 %
MFCD00149180
LVXHNCUCBXIIPE-UHFFFAOYSA-L
disodium phosphoric acid heptahydrate
6096963
98 to 102%
Pass Test
Sodium Phosphate Dibasic Heptahydrate
White
9.0 to 9.3
500 g
H15Na2O11P
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
O.[Na+].[Na+].OP([O-])([O-])=O
268.06
268.07
Certified ACS
Poly Bottle
Videos
Safety and Handling

Safety and Handling

SDS
missing translation for 'documents'

missing translation for 'documents'