missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic dodecahydrate, ≥99% (T), Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Physical Form | Crystalline Powder |
| Quantity | 1 kg |
| Linear Formula | Na2HPO4 · 12H2O |
| Formula Weight | 358.14g/mol |
| Percent Purity | ≥99% (T) |
| Chemical Name or Material | Sodium phosphate dibasic dodecahydrate |
Safety and Handling
| RTECSNumber | TC5725000 |