accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2ONaNaOHPOOO

CAS RN 10039-32-4

IUPAC Name: disodium dodecahydrate hydrogen phosphate
Synonyms: disodium dodecahydrate hydrogen phosphate
Molecular Weight (g/mol): 358.14
Molecular Formula: H25Na2O16P
InChi Key: DGLRDKLJZLEJCY-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O
Molecular Weight (g/mol) 
  • (7)
  • (7)
  • (2)
Quantity 
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (4)
Physical Form 
  • (1)
  • (12)
  • (2)
Grade 
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (5)
  • (2)
  • (7)
  • (7)
  • (7)
  • (2)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (4)
  • (1)
  • (12)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)