missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, dibasic dodecahydrate, 99%, extra pure

Catalog No. p-4813653
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC448160010 1 kg Plastic bottle
AC448160050 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448160010 Supplier Thermo Scientific Chemicals Supplier No. 448160010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10039-32-4 | HNa2O412H2O | 358.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10039-32-4
Molecular Formula HNa2O412H2O
Molecular Weight (g/mol) 358.13
InChI Key DGLRDKLJZLEJCY-UHFFFAOYSA-L
Synonym disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
PubChem CID 61456
IUPAC Name disodium;hydrogen phosphate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]

Specifications

Melting Point 35.0°C
Quantity 1 kg
Assay Percent Range 99%
Linear Formula Na2HPO4·12H2O
Merck Index 15,8789
Solubility Information Solubility in water: 218g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 358.13
Percent Purity 99%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Sodium phosphate, dibasic dodecahydrate
Show More Show Less

RUO – Research Use Only