missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, dibasic dodecahydrate, 98.5%, for analysis

CAS: 10039-32-4 | HNa2O412H2O | 358.13 g/mol

Supplier:  Thermo Scientific Chemicals 271060025

 View more versions of this product

Catalog No. AC271060025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium phosphate, dibasic dodecahydrate
White
9 to 9.4 (5% aq. soln.)
2.5 kg
HNa2O412H2O
MFCD00149181
disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
DGLRDKLJZLEJCY-UHFFFAOYSA-L
disodium;hydrogen phosphate;dodecahydrate
61456
98.5%
Plastic bottle
35.0°C
10039-32-4
Crystals
98.5%
Na2HPO4·12H2O
15, 8789
Solubility in water: 218g/L (20°C). Other solubilities: practically insoluble in alcohol
O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]
358.13
358.13
Analytical

Chemical Identifiers

10039-32-4
358.13
DGLRDKLJZLEJCY-UHFFFAOYSA-L
61456
O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]
HNa2O412H2O
MFCD00149181
disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
disodium;hydrogen phosphate;dodecahydrate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only