Learn More
Sodium Phosphate Dibasic Dihydrate (USP/FCC/EP/BP), Fisher Chemical™
Supplier: Fisher Chemical S472212
Specifications
| Sodium Phosphate Dibasic Dihydrate | |
| 10028-24-7 | |
| 2.5 kg | |
| Na2HPO4·2H2O | |
| disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o | |
| O.O.[Na+].[Na+].OP([O-])([O-])=O | |
| 177.99 | |
| 177.99 | |
| BP/EP/FCC/USP | |
| 18.0 to 22.0% (FCC), 18.5 to 21.5% (USP), 18.5 to 21.5% (USP), 18.0 to 22.0% (FCC), 19.5 to 21.0% (EP/BP), 19.5 to 21.0% (BP), 19.5 to 21.0% (EP) |
| White | |
| Powder/Solid | |
| H5Na2O6P | |
| MFCD00149182 | |
| KDQPSPMLNJTZAL-UHFFFAOYSA-L | |
| disodium dihydrate hydrogen phosphate | |
| 6451167 | |
| 98.0 to 100.5% (USP) | |
| Pass Test (BP), Pass Test (EP), Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC/EP/BP) | |
| Poly Bottle |
Chemical Identifiers
| 10028-24-7 | |
| 177.99 | |
| KDQPSPMLNJTZAL-UHFFFAOYSA-L | |
| 6451167 | |
| O.O.[Na+].[Na+].OP([O-])([O-])=O |
| H5Na2O6P | |
| MFCD00149182 | |
| disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o | |
| disodium dihydrate hydrogen phosphate |
Safety and Handling
CAUTION!
Emergency Overview
Product dust may be irritating to eyes, skin and respiratory system. Wear personal protective equipment. Ensure adequate ventilation. Use personal protective equipment. Avoid contact with skin, eyes and clothing. Do not breathe dust. Rinse with water. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.
NFPA
Health:0
Flammability:0
Instability:0