missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Dihydrate (USP/FCC/EP/BP), Fisher Chemical™

Catalog No. p-34240
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S472500 500 g Poly Bottle
S472212 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S472500 Supplier Thermo Fisher Scientific Supplier No. S472500
Only null left

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
PubChem CID 6451167
IUPAC Name disodium dihydrate hydrogen phosphate
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Color White
Physical Form Powder/Solid
Quantity 500 g
Linear Formula Na2HPO4·2H2O
Formula Weight 177.99
Percent Purity ≥98.0% (FCC)
Grade BP/EP/FCC/USP
Identification Pass Test (BP), Pass Test (EP), Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC/EP/BP)
Loss on Drying 18.0 to 22.0% (FCC), 18.5 to 21.5% (USP), 18.5 to 21.5% (USP), 18.0 to 22.0% (FCC), 19.5 to 21.0% (EP/BP), 19.5 to 21.0% (BP), 19.5 to 21.0% (EP)
Packaging Poly Bottle
Chemical Name or Material Sodium Phosphate Dibasic Dihydrate
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
Product dust may be irritating to eyes, skin and respiratory system. Wear personal protective equipment. Ensure adequate ventilation. Use personal protective equipment. Avoid contact with skin, eyes and clothing. Do not breathe dust. Rinse with water. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:0
Flammability:0
Instability:0