missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Dibasic Anhydrous (USP), Fisher Chemical™
Supplier: Thermo Fisher Scientific FLS37512
Specifications
| Sodium Phosphate Dibasic Anhydrous | |
| White | |
| Powder/Solid | |
| HNa2O4P | |
| disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
| [Na+].[Na+].OP([O-])([O-])=O | |
| 141.96 | |
| CHEBI:34683 | |
| 98.0 to 100.5% (USP) | |
| Pass Test (USP) | |
| Poly Pail |
| 7558-79-4 | |
| 8.7 to 9.3 | |
| 12 kg | |
| MFCD00003496 | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| disodium hydrogen phosphate | |
| 24203 | |
| 141.96 | |
| USP | |
| 5.0% max. (USP) | |
| Decomposes information: > 240°C |
Chemical Identifiers
| 7558-79-4 | |
| 141.96 | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| 24203 | |
| disodium hydrogen phosphate |
| HNa2O4P | |
| MFCD00003496 | |
| disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
| CHEBI:34683 | |
| [Na+].[Na+].OP([O-])([O-])=O |
Safety and Handling
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health:1 Flammability:0 Instability:1
Signal Word: CAUTION!